Learn More
Cayman Chemical 3-Cyano-7-ethoxycoumarin, Purity: ≥95%, CAS: 117620-77-6, 50 mg
SDP
Catalog No. 502936256
Encompass_Preferred
A fluorogenic CYP450 substrate; a substrate for CYP1A1 and CYP1A2; metabolized to a lesser extent by CYP2D6 and CYP2C19; metabolized to 3-cyano-7-hydroxycoumarin and its fluorescence can be used to quantify CYP450 isoform activity
Catalog No. 50-293-6256 Supplier Cayman Chemical Supplier No. 2544150 MG
A fluorogenic CYP450 substrate; a substrate for CYP1A1 and CYP1A2; metabolized to a lesser extent by CYP2D6 and CYP2C19; metabolized to 3-cyano-7-hydroxycoumarin and its fluorescence can be used to quantify CYP450 isoform activity