Learn More
Cayman Chemical 3-Deaza-2'-deoxyadenosine, Purity: ≥98%, CAS: 78582-17-9, 25 mg
SDP
Catalog No. 502894876
Encompass_Preferred
Strongly inhibits lymphocyte-mediated cytolysis with low cytotoxicity when applied at 100 μM; can also be used to evaluate the role of the adenine N3 nitrogen in DNA structure and function
Catalog No. 50-289-4876 Supplier Cayman Chemical Supplier No. 900078625 MG
Strongly inhibits lymphocyte-mediated cytolysis with low cytotoxicity when applied at 100 μM; can also be used to evaluate the role of the adenine N3 nitrogen in DNA structure and function