Learn More
Cayman Chemical 3-Deaza-2-deoxyadenosIn 5mg
SDP
Catalog No. 502892818
Encompass_Preferred
Strongly inhibits lymphocyte-mediated cytolysis with low cytotoxicity when applied at 100 μM; can also be used to evaluate the role of the adenine N3 nitrogen in DNA structure and function
Catalog No. 50-289-2818 Supplier Cayman Chemical Supplier No. 90007865 MG
Strongly inhibits lymphocyte-mediated cytolysis with low cytotoxicity when applied at 100 μM; can also be used to evaluate the role of the adenine N3 nitrogen in DNA structure and function