Learn More
Selleck Chemical LLC 3-deazaadenosine-5mg
SDP
Supplier:  Selleck Chemical LLC S68355MG
Encompass_Preferred
3-Deazaadenosine is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 µM, and has anti-inflammatory, anti-proliferative and anti-HIV activity.
Catalog No. 50-238-9901