Learn More
Medchemexpress LLC HY-W013332A 1mg Medchemexpress, 3-Deazaadenosine (hydrochloride) CAS:86583-19-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW013332A1MG
Encompass_Preferred
Medchemexpress, HY-W013332A 1mg 3-Deazaadenosine (hydrochloride) CAS:86583-19-9 3-Deazaadenosine (hydrochloride) is an inhibitor of S-adenosylhomocysteine hydrolase , with a K i of 3.9 µM; 3-Deazaadenosine has anti-inflammatory, anti-proliferative and anti- HIV activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4581