Learn More
Medchemexpress LLC 3-Deazaadenosine hydrochloride | 86583-19-9 | 99.7% | 302.72 | 1 ML
SDP
Catalog No. 5000454174
Encompass_Preferred
3-Deazaadenosine hydrochloride is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM. It exhibits anti-inflammatory, anti-proliferative, and anti-HIV activity. This product is for research use only and not sold to patients.
  • Inhibits S-adenosylhomocysteine hydrolase
  • Shows anti-HIV effect
  • Inhibits LPS-induced expression of TNF-α mRNA
  • Enhances nuclear translocation of NF-κB
  • Suppresses vascular smooth muscle cell (VSMC) proliferation
Catalog No. 50-004-54174 Supplier Medchemexpress LLC Supplier No. HYW013332A10MM/1ML
3-Deazaadenosine hydrochloride is an inhibitor of S-adenosylhomocysteine hydrolase, with a Ki of 3.9 μM. It exhibits anti-inflammatory, anti-proliferative, and anti-HIV activity. This product is for research use only and not sold to patients.
  • Inhibits S-adenosylhomocysteine hydrolase
  • Shows anti-HIV effect
  • Inhibits LPS-induced expression of TNF-α mRNA
  • Enhances nuclear translocation of NF-κB
  • Suppresses vascular smooth muscle cell (VSMC) proliferation