Learn More
Selleck Chemical LLC 3-deazaneplanocin A (DZNeP) HCl 1mg 120964-45-6 NSC 617989 HCl
SDP
Catalog No. 501366337
Encompass_Preferred
3-deazaneplanocin A HCl (DZNeP, NSC 617989), an analog of adenosine, is a competitive inhibitor of S-adenosylhomocysteine hydrolase with Ki of 50 pM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6337 Supplier Selleck Chemical LLC Supplier No. S71201MG
3-deazaneplanocin A HCl (DZNeP, NSC 617989), an analog of adenosine, is a competitive inhibitor of S-adenosylhomocysteine hydrolase with Ki of 50 pM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.