Learn More
Apexbio Technology LLC 3-Deazaneplanocin A (DZNep) hydrochloride, 1mg. Cas: 120964-45-6 MFCD: MFCD18782644. SAHH and EZH2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A81821
Encompass_Preferred
3-Deazaneplanocin A hydrochloride is an inhibitor of histone methyltransferase. 120964-45-6; MW 298.73; >14.9mg/mL in DMSO; Purity = 98.00%
Catalog No. NC1049761