Learn More
Medchemexpress LLC HY-12186 1mg Medchemexpress, 3-Deazaneplanocin A (hydrochloride) CAS:120964-45-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY121861MG
Encompass_Preferred
Medchemexpress, HY-12186 1mg 3-Deazaneplanocin A (hydrochloride) CAS:120964-45-6 3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8858