Learn More
Cayman Chemical 3-Deazauridine, Purity: ≥98%, CAS: 23205-42-7, 50 mg
SDP
Catalog No. 502995566
Encompass_Preferred
A nucleoside analog; converted intracellularly to 3-deazauridine triphosphate, which inhibits the biosynthesis of CTP; inhibits the growth of L1210 leukemia cells when used at a concentration of 6 µM; dose-dependently reduces mortality in a mouse model of leukemia; enhances the incorporation of decitabine into DNA in leukemia cells when used at a concentration of 20 µM; reduces mortality when used at a dose of 100 or 150 mg/kg and combined with decitabine in a mouse model of leukemia
Catalog No. 50-299-5566 Supplier Cayman Chemical Supplier No. 2312550 MG
A nucleoside analog; converted intracellularly to 3-deazauridine triphosphate, which inhibits the biosynthesis of CTP; inhibits the growth of L1210 leukemia cells when used at a concentration of 6 µM; dose-dependently reduces mortality in a mouse model of leukemia; enhances the incorporation of decitabine into DNA in leukemia cells when used at a concentration of 20 µM; reduces mortality when used at a dose of 100 or 150 mg/kg and combined with decitabine in a mouse model of leukemia