Learn More
Cayman Chemical 3-demethyl Colchicine, Purity: ≥98%, CAS: 7336-33-6, 10 mg
SDP
Catalog No. 502909333
Encompass_Preferred
An active metabolite of colchicine; suppresses antibody production induced by sheep red blood cells in co-culture with isolated mouse spleen cells from 0.1-10 µg/ml; inhibits carrageenan-induced paw edema in rats at 100 µg/animal
Catalog No. 50-290-9333 Supplier Cayman Chemical Supplier No. 3523810 MG
An active metabolite of colchicine; suppresses antibody production induced by sheep red blood cells in co-culture with isolated mouse spleen cells from 0.1-10 µg/ml; inhibits carrageenan-induced paw edema in rats at 100 µg/animal