Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-deoxy-3'-fluoroadenosine | 75059-22-2 | MFCD20134043 | 99.3% | 269.24 | C10H12FN5O3 | 5 MG
SDP

Catalog No. 5000246779
Encompass_Preferred

3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in biomedical research as a nucleoside antimetabolite. It is applied in studies of nucleoside metabolism, inhibition of DNA synthesis, and induction of apoptosis in preclinical oncology and biochemical assays. The compound is supplied as a solid or as ready-to-use solutions for research applications.

  • Purity 99.3% suitable for research applications.
  • White to off-white solid appearance.
  • Molecular weight 269.24 g/mol.
  • Store protected from light at recommended temperatures for stability.
  • Available in milligram quantities and as DMSO solutions for convenience.

Catalog No. 50-002-46779 Supplier Medchemexpress LLC Supplier No. HY1020185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in biomedical research as a nucleoside antimetabolite. It is applied in studies of nucleoside metabolism, inhibition of DNA synthesis, and induction of apoptosis in preclinical oncology and biochemical assays. The compound is supplied as a solid or as ready-to-use solutions for research applications.

  • Purity 99.3% suitable for research applications.
  • White to off-white solid appearance.
  • Molecular weight 269.24 g/mol.
  • Store protected from light at recommended temperatures for stability.
  • Available in milligram quantities and as DMSO solutions for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.