Learn More
Medchemexpress LLC 3'-deoxy-3'-fluoroadenosine | 75059-22-2 | MFCD20134043 | 99.3% | 269.24 g·mol⁻¹ | C10H12FN5O3 | 10 MG
SDP
Catalog No. 5000247583
Encompass_Preferred
3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in biochemical and cellular research to probe nucleoside metabolism and antitumor mechanisms. The compound contains a fluorine substituent at the 3' position of adenosine and has activity that can inhibit DNA synthesis and promote apoptosis in model systems.
  • High purity, typically 99.3%.
  • Chemical formula C10H12FN5O3; molecular weight 269.24 g·mol⁻¹.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • Pack sizes include small research quantities, for example 10 MG.
  • Suitable for studies of DNA synthesis inhibition and apoptotic pathways.
  • Datasheets and analytical files (COA, HNMR, LCMS, SDS) available.
Catalog No. 50-002-47583 Supplier Medchemexpress LLC Supplier No. HY10201810MG
3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in biochemical and cellular research to probe nucleoside metabolism and antitumor mechanisms. The compound contains a fluorine substituent at the 3' position of adenosine and has activity that can inhibit DNA synthesis and promote apoptosis in model systems.
  • High purity, typically 99.3%.
  • Chemical formula C10H12FN5O3; molecular weight 269.24 g·mol⁻¹.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • Pack sizes include small research quantities, for example 10 MG.
  • Suitable for studies of DNA synthesis inhibition and apoptotic pathways.
  • Datasheets and analytical files (COA, HNMR, LCMS, SDS) available.