Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-Deoxy-3'-fluoroadenosine | 75059-22-2 | MFCD20134043 | 99.3% | 269.24 g·mol⁻¹ | C10H12FN5O3 | 100 MG
SDP

Catalog No. 5000247535
Encompass_Preferred

3'-Deoxy-3'-fluoroadenosine is a fluorinated purine nucleoside analogue (C10H12FN5O3, CAS 75059-22-2) used as a research reagent. It functions as a nucleoside antimetabolite that can interfere with DNA synthesis and has reported antiviral and antitumor activity; it is provided as a high-purity solid for laboratory studies.

  • High purity (99.3%).
  • Molecular weight 269.24 g·mol⁻¹.
  • Chemical formula C10H12FN5O3.
  • Used in antiviral and antitumor research as a nucleoside analogue.
  • Available as a 100 mg solid for laboratory use.

Catalog No. 50-002-47535 Supplier Medchemexpress LLC Supplier No. HY102018100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Deoxy-3'-fluoroadenosine is a fluorinated purine nucleoside analogue (C10H12FN5O3, CAS 75059-22-2) used as a research reagent. It functions as a nucleoside antimetabolite that can interfere with DNA synthesis and has reported antiviral and antitumor activity; it is provided as a high-purity solid for laboratory studies.

  • High purity (99.3%).
  • Molecular weight 269.24 g·mol⁻¹.
  • Chemical formula C10H12FN5O3.
  • Used in antiviral and antitumor research as a nucleoside analogue.
  • Available as a 100 mg solid for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.