Learn More
Medchemexpress LLC 3'-deoxy-3'-fluoroadenosine | 75059-22-2 | MFCD20134043 | 99.3% | 269.24 g/mol | C10H12FN5O3 | 50 MG
SDP
Catalog No. 5000246794
Encompass_Preferred
3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in research to investigate nucleoside metabolism and antitumor mechanisms; it is supplied as a solid powder and as a DMSO solution and is characterized by defined molecular properties and high purity.
  • High purity solid suitable for research applications.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • Useful for studies of DNA synthesis inhibition and apoptosis.
  • Molecular formula C10H12FN5O3; molecular weight 269.24 g/mol.
  • Supplied in multiple pack sizes to support small- and medium-scale experiments.
Catalog No. 50-002-46794 Supplier Medchemexpress LLC Supplier No. HY10201850MG
3'-Deoxy-3'-fluoroadenosine is a purine nucleoside analogue used in research to investigate nucleoside metabolism and antitumor mechanisms; it is supplied as a solid powder and as a DMSO solution and is characterized by defined molecular properties and high purity.
  • High purity solid suitable for research applications.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • Useful for studies of DNA synthesis inhibition and apoptosis.
  • Molecular formula C10H12FN5O3; molecular weight 269.24 g/mol.
  • Supplied in multiple pack sizes to support small- and medium-scale experiments.