Learn More
Medchemexpress LLC 3′-deoxy-3′-fluoroguanosine | 123402-21-1 | MFCD09263707 | >98.0% | 285.23 g/mol | C10H12FN5O4 | 100 MG
SDP
Catalog No. 5000282838
Encompass_Preferred
3'-Deoxy-3'-fluoroguanosine is a fluorinated deoxyguanosine nucleoside analogue used in antiviral and biochemical research. It is supplied as a high-purity solid suitable for in vitro studies, enzymatic assays, and compound screening.
  • High purity (>98%).
  • Verified molecular formula and weight (C10H12FN5O4; 285.23 g/mol).
  • Available in common laboratory pack formats, including 100 mg and solution aliquots.
  • Suitable for antiviral, enzymatic, and pharmacological assays.
  • Documented CAS and database identifiers for unambiguous reference.
Catalog No. 50-002-82838 Supplier Medchemexpress LLC Supplier No. HYW128400100MG
3'-Deoxy-3'-fluoroguanosine is a fluorinated deoxyguanosine nucleoside analogue used in antiviral and biochemical research. It is supplied as a high-purity solid suitable for in vitro studies, enzymatic assays, and compound screening.
  • High purity (>98%).
  • Verified molecular formula and weight (C10H12FN5O4; 285.23 g/mol).
  • Available in common laboratory pack formats, including 100 mg and solution aliquots.
  • Suitable for antiviral, enzymatic, and pharmacological assays.
  • Documented CAS and database identifiers for unambiguous reference.