Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3′-deoxy-3′-fluoroguanosine | 123402-21-1 | MFCD09263707 | >98.0% | 285.23 g/mol | C10H12FN5O4 | 25 MG
SDP

Catalog No. 5000282731
Encompass_Preferred

3′-Deoxy-3′-fluoroguanosine is a fluorinated guanosine nucleoside analogue used in biochemical and antiviral research. It is employed to study viral RNA synthesis and polymerase interactions and is supplied as a high-purity solid suitable for laboratory assays.

  • Fluorinated guanosine nucleoside analogue
  • Used to probe viral RNA synthesis and polymerase activity
  • Molecular formula C10H12FN5O4
  • Molecular weight 285.23 g/mol
  • CAS number 123402-21-1
  • Purity >98.0%
  • Supplied as a solid (powder) for research use
  • Also available as concentrated solution preparations for in vitro assays

Catalog No. 50-002-82731 Supplier Medchemexpress LLC Supplier No. HYW12840025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-Deoxy-3′-fluoroguanosine is a fluorinated guanosine nucleoside analogue used in biochemical and antiviral research. It is employed to study viral RNA synthesis and polymerase interactions and is supplied as a high-purity solid suitable for laboratory assays.

  • Fluorinated guanosine nucleoside analogue
  • Used to probe viral RNA synthesis and polymerase activity
  • Molecular formula C10H12FN5O4
  • Molecular weight 285.23 g/mol
  • CAS number 123402-21-1
  • Purity >98.0%
  • Supplied as a solid (powder) for research use
  • Also available as concentrated solution preparations for in vitro assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.