Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3′-deoxy-3′-fluoroguanosine | 123402-21-1 | MFCD09263707 | >98.0% | 285.24 g/mol | C10H12FN5O4 | 10 MG
SDP

Catalog No. 5000282935
Encompass_Preferred

3′-Deoxy-3′-fluoroguanosine is a fluorinated guanosine nucleoside analog supplied for laboratory research. It is used in nucleic acid and antiviral studies where modified nucleosides are required to probe polymerase activity and viral replication. The material is provided as a solid or solution format for experimental assays.

  • Fluorinated guanosine nucleoside analog for biochemical research.
  • Exhibits antiviral activity against certain RNA viruses via RNA-dependent RNA polymerase interaction.
  • Purity greater than 98% as measured by supplier quality control.
  • Molecular formula C10H12FN5O4; molecular weight 285.24 g/mol.
  • Available in small-scale research formats including 10 mg and 100 mg, and 10 mM solution.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-82935 Supplier Medchemexpress LLC Supplier No. HYW12840010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-Deoxy-3′-fluoroguanosine is a fluorinated guanosine nucleoside analog supplied for laboratory research. It is used in nucleic acid and antiviral studies where modified nucleosides are required to probe polymerase activity and viral replication. The material is provided as a solid or solution format for experimental assays.

  • Fluorinated guanosine nucleoside analog for biochemical research.
  • Exhibits antiviral activity against certain RNA viruses via RNA-dependent RNA polymerase interaction.
  • Purity greater than 98% as measured by supplier quality control.
  • Molecular formula C10H12FN5O4; molecular weight 285.24 g/mol.
  • Available in small-scale research formats including 10 mg and 100 mg, and 10 mM solution.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.