Learn More
Medchemexpress LLC 3'-deoxy-3'-fluoroguanosine | 123402-21-1 | MFCD09263707 | 99.3% | 285.23 g/mol | C10H12FN5O4 | 5 MG
SDP
Catalog No. 5000282729
Encompass_Preferred
3'-Deoxy-3'-fluoroguanosine is a nucleoside analogue research compound reported to exhibit antiviral activity and intended for in vitro antiviral and nucleic acid metabolism studies. It is supplied in small quantities for laboratory assay development and biochemical research.
  • Nucleoside analogue suitable for antiviral research.
  • Reported antiviral activity against tick-borne encephalitis virus.
  • High purity (~99.3%) as indicated in batch documentation.
  • Provided in small research quantities for assay development.
Catalog No. 50-002-82729 Supplier Medchemexpress LLC Supplier No. HYW1284005MG
3'-Deoxy-3'-fluoroguanosine is a nucleoside analogue research compound reported to exhibit antiviral activity and intended for in vitro antiviral and nucleic acid metabolism studies. It is supplied in small quantities for laboratory assay development and biochemical research.
  • Nucleoside analogue suitable for antiviral research.
  • Reported antiviral activity against tick-borne encephalitis virus.
  • High purity (~99.3%) as indicated in batch documentation.
  • Provided in small research quantities for assay development.