Learn More
Medchemexpress LLC 3'-deoxy-3'-fluoroguanosine | 123402-21-1 | MFCD09263707 | 99.3% | 285.23 g/mol | C10H12FN5O4 | 50 MG
SDP
Catalog No. 5000283671
Encompass_Preferred
3'-Deoxy-3'-fluoroguanosine is a fluorinated guanosine analog used as a research reagent in antiviral studies and biochemical assays. It is employed as a probe of nucleoside metabolism and viral polymerase activity, supplied as a high-purity solid for laboratory use.
  • Fluorinated at the 3' position for altered metabolic properties.
  • Useful as an antiviral research reagent and polymerase probe.
  • High purity suitable for biochemical and analytical assays.
  • Supplied as a solid for convenient handling and storage.
  • Molecular formula C10H12FN5O4; molecular weight 285.23 g/mol.
Catalog No. 50-002-83671 Supplier Medchemexpress LLC Supplier No. HYW12840050MG
3'-Deoxy-3'-fluoroguanosine is a fluorinated guanosine analog used as a research reagent in antiviral studies and biochemical assays. It is employed as a probe of nucleoside metabolism and viral polymerase activity, supplied as a high-purity solid for laboratory use.
  • Fluorinated at the 3' position for altered metabolic properties.
  • Useful as an antiviral research reagent and polymerase probe.
  • High purity suitable for biochemical and analytical assays.
  • Supplied as a solid for convenient handling and storage.
  • Molecular formula C10H12FN5O4; molecular weight 285.23 g/mol.