Learn More
Cayman Chemical 3'-Deoxy-3'-fluoroguanosine, Purity: ≥98%, CAS: 123402-21-1, 250 mg
SDP
Catalog No. 502913004
Encompass_Preferred
A nucleoside analog an inhibitor of HCV NS5B (IC50 = 1.8 µM)
Catalog No. 50-291-3004 Supplier Cayman Chemical Supplier No. 35699250 MG
A nucleoside analog an inhibitor of HCV NS5B (IC50 = 1.8 µM)