Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-deoxy-3'-fluorouridine | 57944-13-5 | MFCD02683987 | >95.0% | 246.19 g/mol | C9H11FN2O5 | 10 MG
SDP

Catalog No. 5000286607
Encompass_Preferred

3'-Deoxy-3'-fluorouridine is a fluorinated uridine analogue used in biochemical and oncology research as a probe of nucleoside metabolism and as a research compound in antitumor studies. It is supplied as an off-white powder for laboratory use; consult the product SDS for handling and storage requirements.

  • Chemical name: 3'-deoxy-3'-fluorouridine.
  • CAS number 57944-13-5.
  • Molecular formula C9H11FN2O5.
  • Molecular weight 246.19 g/mol.
  • Purity >95.0%.
  • Appearance off-white to white powder.
  • Storage recommended -20 °C, dry and protected from moisture.
  • Typical packaging 10 mg vial.

Catalog No. 50-002-86607 Supplier Medchemexpress LLC Supplier No. HY15276510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-Deoxy-3'-fluorouridine is a fluorinated uridine analogue used in biochemical and oncology research as a probe of nucleoside metabolism and as a research compound in antitumor studies. It is supplied as an off-white powder for laboratory use; consult the product SDS for handling and storage requirements.

  • Chemical name: 3'-deoxy-3'-fluorouridine.
  • CAS number 57944-13-5.
  • Molecular formula C9H11FN2O5.
  • Molecular weight 246.19 g/mol.
  • Purity >95.0%.
  • Appearance off-white to white powder.
  • Storage recommended -20 °C, dry and protected from moisture.
  • Typical packaging 10 mg vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.