Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) | 153666-19-4 | 99.9% | C9H16O9 | 25 MG
SDP

Catalog No. 5000356617
Encompass_Preferred

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a type of sialic acid that can be hydrolyzed by sialidase from the loach (Misgurnus fossilis).

  • Solid appearance
  • White to off-white color
  • Soluble in water at 100 mg/mL (372.83 mM), requires ultrasonic
  • Purity of 99.9%
  • Molecular weight of 268.22
  • Chemical formula: C9H16O9
  • CAS number: 153666-19-4
  • Ships at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-56617 Supplier Medchemexpress LLC Supplier No. 25MGHY145533
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a type of sialic acid that can be hydrolyzed by sialidase from the loach (Misgurnus fossilis).

  • Solid appearance
  • White to off-white color
  • Soluble in water at 100 mg/mL (372.83 mM), requires ultrasonic
  • Purity of 99.9%
  • Molecular weight of 268.22
  • Chemical formula: C9H16O9
  • CAS number: 153666-19-4
  • Ships at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.