Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) | 153666-19-4 | 99.9% | C9H16O9 | 50 MG
SDP

Catalog No. 5000356011
Encompass_Preferred

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a type of sialic acid. It can be hydrolyzed by sialidase from the loach (Misgurnus fossilis). This product is supplied as a white to off-white solid.

  • High purity: 99.90%
  • Molecular weight: 268.22
  • Solubility in water: 100 mg/mL
  • Stable as powder at -20°C for 3 years, 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-56011 Supplier Medchemexpress LLC Supplier No. HY14553350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a type of sialic acid. It can be hydrolyzed by sialidase from the loach (Misgurnus fossilis). This product is supplied as a white to off-white solid.

  • High purity: 99.90%
  • Molecular weight: 268.22
  • Solubility in water: 100 mg/mL
  • Stable as powder at -20°C for 3 years, 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.