Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-deoxy-d-glycero-d-galacto-2-nonulosonic acid (KDN) | 153666-19-4 | MFCD00269963 | 99.9% | 268.22 g/mol | C9H16O9 | 5 MG
SDP

Catalog No. 5000221070
Encompass_Preferred

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a sialic acid derivative provided as a high-purity research reagent. Supplied as a white to off-white solid, it is intended for glycobiology, carbohydrate analysis, and biochemical assays. Documentation such as a certificate of analysis and safety data sheets are available; follow recommended storage to maintain stability.

  • High purity suitable for analytical and research use (99.9%).
  • Available in milligram-scale packages for small-scale experiments.
  • Molecular weight 268.22 g/mol and formula C9H16O9 for stoichiometric calculations.
  • White to off-white solid, easy to weigh and dissolve for assays.
  • Certificate of analysis and safety data sheets available from the supplier.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-21070 Supplier Medchemexpress LLC Supplier No. HY1455335MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a sialic acid derivative provided as a high-purity research reagent. Supplied as a white to off-white solid, it is intended for glycobiology, carbohydrate analysis, and biochemical assays. Documentation such as a certificate of analysis and safety data sheets are available; follow recommended storage to maintain stability.

  • High purity suitable for analytical and research use (99.9%).
  • Available in milligram-scale packages for small-scale experiments.
  • Molecular weight 268.22 g/mol and formula C9H16O9 for stoichiometric calculations.
  • White to off-white solid, easy to weigh and dissolve for assays.
  • Certificate of analysis and safety data sheets available from the supplier.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.