Learn More
Medchemexpress LLC 3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (sodium) | 95.0% | C9H15NaO9 | 25 MG
SDP
Catalog No. 5000354061
Encompass_Preferred
3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) sodium is a sialic acid that protects oligo/(poly)sialyl chains from exosialidases at the nonreducing terminal. It is abundant in fetal cord red blood cells and malignant human ovarian cancer cells. This compound also plays a role in the egg activation of salmonid fish.
  • Sialic acid compound
  • Protects oligo/(poly)sialyl chains
  • Abundant in fetal cord red blood cells
  • Abundant in malignant human ovarian cancer cells
  • Plays a role in egg activation of salmonid fish
Catalog No. 50-003-54061 Supplier Medchemexpress LLC Supplier No. 25MGHY145533A
3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) sodium is a sialic acid that protects oligo/(poly)sialyl chains from exosialidases at the nonreducing terminal. It is abundant in fetal cord red blood cells and malignant human ovarian cancer cells. This compound also plays a role in the egg activation of salmonid fish.
  • Sialic acid compound
  • Protects oligo/(poly)sialyl chains
  • Abundant in fetal cord red blood cells
  • Abundant in malignant human ovarian cancer cells
  • Plays a role in egg activation of salmonid fish