Learn More
Medchemexpress LLC 3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid sodium | 95.0% | C9H15NaO9 | 50 MG
SDP
Catalog No. 5000354447
Encompass_Preferred
3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) sodium is a sialic acid. It protects the oligo/(poly)sialyl chains from exosialidases at the nonreducing terminal and plays a role in egg activation of salmonid fish. This compound is abundant in fetal cord red blood cells and malignant human ovarian cancer cells.
  • Sialic acid
  • Protects oligo/(poly)sialyl chains from exosialidases
  • Plays a role in egg activation of salmonid fish
  • Abundant in fetal cord red blood cells
  • Abundant in malignant human ovarian cancer cells
Catalog No. 50-003-54447 Supplier Medchemexpress LLC Supplier No. HY145533A50MG
3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) sodium is a sialic acid. It protects the oligo/(poly)sialyl chains from exosialidases at the nonreducing terminal and plays a role in egg activation of salmonid fish. This compound is abundant in fetal cord red blood cells and malignant human ovarian cancer cells.
  • Sialic acid
  • Protects oligo/(poly)sialyl chains from exosialidases
  • Plays a role in egg activation of salmonid fish
  • Abundant in fetal cord red blood cells
  • Abundant in malignant human ovarian cancer cells