Learn More
Medchemexpress LLC 3-deoxy-galactosone | 4134-97-8 | MFCD18643020 | 162.14 g/mol | C6H10O5 | 5 MG
SDP
Catalog No. 5000229005
Encompass_Preferred
3-Deoxy-galactosone (CAS 4134-97-8) is an endogenous 1,2-dicarbonyl compound produced by degradation of galactose and formed in foods during Maillard and caramelization reactions. It can be generated from 3-deoxyglucosone (3-DG) via the intermediate 3,4-dideoxyglucosone-3-ene (3,4-DGE). Molecular formula C6H10O5; molecular weight 162.14 g/mol. Purity and appearance are on the product page - consult the Certificate of Analysis for lot-specific data.
  • Endogenous 1,2-dicarbonyl metabolite relevant to food chemistry and metabolic studies.
  • Provided in small research pack sizes suitable for analytical and reference use.
  • Molecular formula C6H10O5 and molecular weight 162.14 g/mol support identification and calculations.
  • Consult Certificate of Analysis for lot-specific purity and storage instructions.
Catalog No. 50-002-29005 Supplier Medchemexpress LLC Supplier No. HY1188135MG
3-Deoxy-galactosone (CAS 4134-97-8) is an endogenous 1,2-dicarbonyl compound produced by degradation of galactose and formed in foods during Maillard and caramelization reactions. It can be generated from 3-deoxyglucosone (3-DG) via the intermediate 3,4-dideoxyglucosone-3-ene (3,4-DGE). Molecular formula C6H10O5; molecular weight 162.14 g/mol. Purity and appearance are on the product page - consult the Certificate of Analysis for lot-specific data.
  • Endogenous 1,2-dicarbonyl metabolite relevant to food chemistry and metabolic studies.
  • Provided in small research pack sizes suitable for analytical and reference use.
  • Molecular formula C6H10O5 and molecular weight 162.14 g/mol support identification and calculations.
  • Consult Certificate of Analysis for lot-specific purity and storage instructions.