Learn More
Cayman Chemical 3-deoxy Glucosone, Purity: ≥95%, CAS: 4084-27-9, 10 mg
SDP
Catalog No. 502972062
Encompass_Preferred
A highly reactive 2-oxoaldehyde intermediate of the Maillard reaction produced during oxidative stress in response to excess sugar consumption and in association with diabetes; glycates and crosslinks proteins leading to the formation of advanced glycation endproducts
Catalog No. 50-297-2062 Supplier Cayman Chemical Supplier No. 1634710 MG
A highly reactive 2-oxoaldehyde intermediate of the Maillard reaction produced during oxidative stress in response to excess sugar consumption and in association with diabetes; glycates and crosslinks proteins leading to the formation of advanced glycation endproducts