Learn More
Cayman Chemical 3′-deoxy Thymidine, Purity: ≥98%, CAS: 3416-05-5, 50 mg
SDP
Catalog No. 502947999
Encompass_Preferred
An antiviral nucleoside analog; inhibits HIV replication in infected human PBMCs (EC50 = 0.17 µM); reduces viral plaque formation in HSV-1-infected CV-1 cells at 1, 3, 10, or 30 µM
Catalog No. 50-294-7999 Supplier Cayman Chemical Supplier No. 3393950 MG
An antiviral nucleoside analog; inhibits HIV replication in infected human PBMCs (EC50 = 0.17 µM); reduces viral plaque formation in HSV-1-infected CV-1 cells at 1, 3, 10, or 30 µM