Learn More
AdipoGen 3-Deoxyaphidicolin
SDP
Supplier:  AdipoGen BVT0451C250
Encompass_Preferred
Chemical. CAS 85483-00-7. Formula C20H34O3. MW 322.5. Isolated from Phoma sp. BS 7210. Inhibitor of eukaryotic DNA synthesis. Mycotoxin. Phytotoxic and cytotoxic agent. Specific inhibitor of DNA polymerase alpha.
Catalog No. 50-256-1912