Learn More
Medchemexpress LLC 3'-Deoxycytidine | 7057-33-2 | 98.0% | 227.22 | 10 MG
SDP
Catalog No. 5000452799
Encompass_Preferred
3'-Deoxycytidine is a purine nucleoside analogue (4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one) with broad antitumor activity against indolent lymphoid malignancies. Its anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.
  • Inhibits replication of S180 cell line in vitro with an ED50 of 15 μM.
  • Inhibits replication of CCRF-CEM cell line in vitro with an ED50 of 25 μM.
  • Inhibits replication of L1210 cell line in vitro with an ED50 of 5 μM.
  • Inhibits replication of P388 cell line in vitro with an ED50 of 2.5 μM.
Catalog No. 50-004-52799 Supplier Medchemexpress LLC Supplier No. HYW01229310MG
3'-Deoxycytidine is a purine nucleoside analogue (4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one) with broad antitumor activity against indolent lymphoid malignancies. Its anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.
  • Inhibits replication of S180 cell line in vitro with an ED50 of 15 μM.
  • Inhibits replication of CCRF-CEM cell line in vitro with an ED50 of 25 μM.
  • Inhibits replication of L1210 cell line in vitro with an ED50 of 5 μM.
  • Inhibits replication of P388 cell line in vitro with an ED50 of 2.5 μM.