Learn More
Medchemexpress LLC 3'-Deoxycytidine | 7057-33-2 | 98.0% | 227.22 | 5 MG
SDP
Catalog No. 5000453659
Encompass_Preferred
3'-Deoxycytidine (4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one) is a purine nucleoside analogue. These analogues demonstrate broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms primarily involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analogue
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Anticancer mechanisms involve inhibition of DNA synthesis
  • Induction of apoptosis
Catalog No. 50-004-53659 Supplier Medchemexpress LLC Supplier No. HYW0122935MG
3'-Deoxycytidine (4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one) is a purine nucleoside analogue. These analogues demonstrate broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms primarily involve the inhibition of DNA synthesis and the induction of apoptosis.
  • Purine nucleoside analogue
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Anticancer mechanisms involve inhibition of DNA synthesis
  • Induction of apoptosis