Learn More
Medchemexpress LLC 3'-Deoxycytidine | 7057-33-2 | 98.0% | 227.22 | 50 MG
SDP
Catalog No. 5000452828
Encompass_Preferred
4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one (3'-Deoxycytidine) is a purine nucleoside analogue. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Functions as a purine nucleoside analogue.
  • Demonstrates broad antitumor activity.
  • Targets indolent lymphoid malignancies.
  • Inhibits DNA synthesis.
  • Induces apoptosis.
Catalog No. 50-004-52828 Supplier Medchemexpress LLC Supplier No. HYW01229350MG
4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one (3'-Deoxycytidine) is a purine nucleoside analogue. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Functions as a purine nucleoside analogue.
  • Demonstrates broad antitumor activity.
  • Targets indolent lymphoid malignancies.
  • Inhibits DNA synthesis.
  • Induces apoptosis.