Learn More
Medchemexpress LLC 3'-Deoxycytidine | 7057-33-2 | MFCD00079144 | 98.0% | 227.22 | 1 ML
SDP
Catalog No. 5000452827
Encompass_Preferred
4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one (3'-Deoxycytidine) is a purine nucleoside analogue. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Purine nucleoside analogue.
  • Has broad antitumor activity targeting indolent lymphoid malignancies.
  • Anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.
Catalog No. 50-004-52827 Supplier Medchemexpress LLC Supplier No. HYW01229310MM/1ML
4-Amino-1-((2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one (3'-Deoxycytidine) is a purine nucleoside analogue. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Purine nucleoside analogue.
  • Has broad antitumor activity targeting indolent lymphoid malignancies.
  • Anticancer mechanisms involve inhibition of DNA synthesis and induction of apoptosis.