Learn More
Medchemexpress LLC 3′-Deoxyguanosine | 3608-58-0 | 97.0% | 267.25 | 10 MG
SDP
Catalog No. 5000454425
Encompass_Preferred
3′-Deoxyguanosine is a derivative of Guanosine that interacts with human purine nucleoside phosphorylase through hydrogen bonding. It demonstrates moderate displacement activity for [3H]-guanosine in rat meninges. This product is for research use only and not sold to patients.
  • Derivative of guanosine
  • Interacts with human purine nucleoside phosphorylase
  • Useful for studying guanosine receptors
  • Useful for studying purine metabolism
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges
Catalog No. 50-004-54425 Supplier Medchemexpress LLC Supplier No. HYW01333010MG
3′-Deoxyguanosine is a derivative of Guanosine that interacts with human purine nucleoside phosphorylase through hydrogen bonding. It demonstrates moderate displacement activity for [3H]-guanosine in rat meninges. This product is for research use only and not sold to patients.
  • Derivative of guanosine
  • Interacts with human purine nucleoside phosphorylase
  • Useful for studying guanosine receptors
  • Useful for studying purine metabolism
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges