Learn More
Medchemexpress LLC 3′-Deoxyguanosine | 3608-58-0 | 97.0% | 267.25 | 25 MG
SDP
Catalog No. 5000453203
Encompass_Preferred
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase through hydrogen bonding with residues like Glu201 and Asn243. This compound demonstrates moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Derivative of guanosine.
  • Interacts with human purine nucleoside phosphorylase via hydrogen bonding.
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges.
  • Useful for studying guanosine receptor mechanisms.
  • Useful for studying purine metabolism.
  • Displaces 50 nM [3H]-guanosine binding to rat brain membranes.
Catalog No. 50-004-53203 Supplier Medchemexpress LLC Supplier No. 25MGHYW013330
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase through hydrogen bonding with residues like Glu201 and Asn243. This compound demonstrates moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Derivative of guanosine.
  • Interacts with human purine nucleoside phosphorylase via hydrogen bonding.
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges.
  • Useful for studying guanosine receptor mechanisms.
  • Useful for studying purine metabolism.
  • Displaces 50 nM [3H]-guanosine binding to rat brain membranes.