Learn More
Medchemexpress LLC 3′-Deoxyguanosine | 3608-58-0 | 97.0% | 267.25 | 5 MG
SDP
Catalog No. 5000454401
Encompass_Preferred
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase through hydrogen bonding with residues like Glu201 and Asn243. This compound shows moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Derivative of Guanosine.
  • Interacts with human purine nucleoside phosphorylase via hydrogen bonding.
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges.
  • Useful for studying guanosine receptor mechanisms and purine metabolism.
  • For research use only.
Catalog No. 50-004-54401 Supplier Medchemexpress LLC Supplier No. HYW0133305MG
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase through hydrogen bonding with residues like Glu201 and Asn243. This compound shows moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Derivative of Guanosine.
  • Interacts with human purine nucleoside phosphorylase via hydrogen bonding.
  • Exhibits moderate displacement activity for [3H]-guanosine in rat meninges.
  • Useful for studying guanosine receptor mechanisms and purine metabolism.
  • For research use only.