Learn More
Medchemexpress LLC 3′-Deoxyguanosine | 3608-58-0 | MFCD28041154 | 97.0% | 267.25 | 100 MG
SDP
Catalog No. 5000454601
Encompass_Preferred
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase via hydrogen bonding with residues such as Glu201 and Asn243. It exhibits moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Useful for studying guanosine receptors
  • Used in research on purine metabolism
  • Interacts via hydrogen bonding with human purine nucleoside phosphorylase
Catalog No. 50-004-54601 Supplier Medchemexpress LLC Supplier No. HYW013330100MG
3′-Deoxyguanosine is a derivative of Guanosine. It interacts with human purine nucleoside phosphorylase via hydrogen bonding with residues such as Glu201 and Asn243. It exhibits moderate displacement activity for [3H]-guanosine in rat meninges and is useful for studying the mechanism of guanosine receptors and purine metabolism.
  • Useful for studying guanosine receptors
  • Used in research on purine metabolism
  • Interacts via hydrogen bonding with human purine nucleoside phosphorylase