Learn More
Cayman Chemical 3-DeoxygunosIn 50mg
SDP
Catalog No. 502968369
Encompass_Preferred
A ligand that can be complexed with enzymes, such as purine nucleoside phosphorylase, and receptors, such as G-coupled receptors, in order to study structure-activity relationships; also used to selectively impair transcription
Catalog No. 50-296-8369 Supplier Cayman Chemical Supplier No. 1790750 MG
A ligand that can be complexed with enzymes, such as purine nucleoside phosphorylase, and receptors, such as G-coupled receptors, in order to study structure-activity relationships; also used to selectively impair transcription