Learn More
Medchemexpress LLC 3'-deoxyuridine-5'-triphosphate trisodium | 00-00-0 | 98.1% | 534.09 g·mol⁻¹ | C9H12N2Na3O14P3 | 5 MG
SDP
Catalog No. 5000222035
Encompass_Preferred
3'-Deoxyuridine-5'-triphosphate trisodium is a deoxyuridine nucleoside triphosphate analog supplied as the trisodium salt for use in molecular biology and enzymology applications. It is provided as a solid reagent with characterized purity and analytical documentation for research use.
  • Supplied as the trisodium salt, suitable for biochemical assays.
  • High purity: approximately 98.1% by reported analysis.
  • Solid, off-white to light yellow appearance for easy handling.
  • Documented analytical data available (COA, NMR, LC-MS, SDS).
  • Recommended storage: store sealed, away from moisture and light; -20°C for solids.
  • Available in small research pack sizes (for example, 5 MG).
Catalog No. 50-002-22035 Supplier Medchemexpress LLC Supplier No. HY135780A5MG
3'-Deoxyuridine-5'-triphosphate trisodium is a deoxyuridine nucleoside triphosphate analog supplied as the trisodium salt for use in molecular biology and enzymology applications. It is provided as a solid reagent with characterized purity and analytical documentation for research use.
  • Supplied as the trisodium salt, suitable for biochemical assays.
  • High purity: approximately 98.1% by reported analysis.
  • Solid, off-white to light yellow appearance for easy handling.
  • Documented analytical data available (COA, NMR, LC-MS, SDS).
  • Recommended storage: store sealed, away from moisture and light; -20°C for solids.
  • Available in small research pack sizes (for example, 5 MG).