Learn More
Medchemexpress LLC 3′-Deoxyuridine | 7057-27-4 | 98.0% | 228.20 | 1 ML
SDP
Catalog No. 5000452825
Encompass_Preferred
3′-Deoxyuridine is a potential anticancer and antiviral agent. It has been shown to inhibit bovine diarrhea virus (BVDV) production.
  • Potential anticancer agent.
  • Potential antiviral agent, inhibiting bovine diarrhea virus (BVDV) production.
Catalog No. 50-004-52825 Supplier Medchemexpress LLC Supplier No. HYW01228210MM/1ML
3′-Deoxyuridine is a potential anticancer and antiviral agent. It has been shown to inhibit bovine diarrhea virus (BVDV) production.
  • Potential anticancer agent.
  • Potential antiviral agent, inhibiting bovine diarrhea virus (BVDV) production.