Learn More
Medchemexpress LLC 3′-Deoxyuridine | 7057-27-4 | 98.0% | 228.20 | 25 MG
SDP
Catalog No. 5000452128
Encompass_Preferred
3′-Deoxyuridine is a potential anticancer and antiviral agent that inhibits bovine diarrhoea virus (BVDV) production. It is for research use only and appears as a white to light yellow solid.
  • Potential anticancer agent
  • Potential antiviral agent
  • Inhibits bovine diarrhoea virus (BVDV) production
  • For research use only
  • Appearance: Solid
  • Color: White to light yellow
Catalog No. 50-004-52128 Supplier Medchemexpress LLC Supplier No. 25MGHYW012282
3′-Deoxyuridine is a potential anticancer and antiviral agent that inhibits bovine diarrhoea virus (BVDV) production. It is for research use only and appears as a white to light yellow solid.
  • Potential anticancer agent
  • Potential antiviral agent
  • Inhibits bovine diarrhoea virus (BVDV) production
  • For research use only
  • Appearance: Solid
  • Color: White to light yellow