Learn More
Cayman Chemical 3-Dephosphocoenzyme A 10mg
SDP
Catalog No. 502939840
Encompass_Preferred
An intermediate in the biosynthesis of CoA from pantothenic acid; phosphorylated by CoA synthase in humans to form CoA; can be used as a transcription initiator in the synthesis of CoA-RNA by in vitro transcription
Catalog No. 50-293-9840 Supplier Cayman Chemical Supplier No. 2739010 MG
An intermediate in the biosynthesis of CoA from pantothenic acid; phosphorylated by CoA synthase in humans to form CoA; can be used as a transcription initiator in the synthesis of CoA-RNA by in vitro transcription