Learn More
Medchemexpress LLC 3'-dephosphocoenzyme A | 3633-59-8 | 96.0% | 687.55 g/mol | C21H35N7O13P2S | 5 MG
SDP
Catalog No. 5000222975
Encompass_Preferred
Dephospho-CoA is a biochemical precursor in coenzyme A biosynthesis supplied as a research reagent. It undergoes the final phosphorylation step to form CoA and is used in studies of cellular metabolism, mitochondrial function, and cell proliferation. Provided as a small-scale, high-purity solid for laboratory research.
Catalog No. 50-002-22975 Supplier Medchemexpress LLC Supplier No. HY1378995MG
Dephospho-CoA is a biochemical precursor in coenzyme A biosynthesis supplied as a research reagent. It undergoes the final phosphorylation step to form CoA and is used in studies of cellular metabolism, mitochondrial function, and cell proliferation. Provided as a small-scale, high-purity solid for laboratory research.