Learn More
New England Biolabs, Inc. 3'-Desthiobiotin-GTP – 0.5 µmol
SDP
Catalog No. 501526417
Encompass
Supplied in: Ultrapure Water as a Triethylammonium salt at pH 7.0 Molecular Formula: C31H52N11O19P3 Molecular Weight: 975.74 g/mol Extinction Coefficient: 260 = ~11,700 Lmol-1 cm-1
Catalog No. 50-152-6417 Supplier New England Biolabs, Inc. Supplier No. N0761S
Supplied in: Ultrapure Water as a Triethylammonium salt at pH 7.0 Molecular Formula: C31H52N11O19P3 Molecular Weight: 975.74 g/mol Extinction Coefficient: 260 = ~11,700 Lmol-1 cm-1