Learn More
Cole-Parmer 3"-Diameter Sieve, Full Height Stainless Steel, No. 70 (212)
Catalog No. NC1698306 Shop All Cole-Parmer Products
Encompass_Preferred
For use with Cole-Parmer sieve shaker (59986-01), sold separately. Order optional stainless steel cover (59988-03) and full-height receiving pan (59988-07), sold separately.
Catalog No. NC1698306 Supplier Cole-Parmer Supplier No. 5998720
For use with Cole-Parmer sieve shaker (59986-01), sold separately. Order optional stainless steel cover (59988-03) and full-height receiving pan (59988-07), sold separately.