Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[Diethoxy(methyl)silyl]propyl Methacrylate 98.0+%, TCI America™
SDP

Catalog No. D467925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D467925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D467925ML Supplier TCI America Supplier No. D467925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65100-04-1
Molecular Formula C12H24O4Si
Molecular Weight (g/mol) 260.405
MDL Number MFCD00053879
InChI Key DOYKFSOCSXVQAN-UHFFFAOYSA-N
Synonym Methacrylic Acid 3-[Diethoxy(methyl)silyl]propyl Ester
PubChem CID 2759529
IUPAC Name 3-[diethoxy(methyl)silyl]propyl 2-methylprop-2-enoate
SMILES CCO[Si](C)(CCCOC(=O)C(=C)C)OCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 260.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[Diethoxy(methyl)silyl]propyl Methacrylate
EINECSNumber (2)-2075
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.