Learn More
Medchemexpress LLC 3-Diethylamino-1-propanol | 622-93-5 | 98.0% | 131.22 | 100 G
SDP
Catalog No. 5000451859
Encompass_Preferred
3-Diethylamino-1-propanol is a tertiary amine compound that demonstrates anticonvulsant activity. It is intended for research use only. The product is supplied as a colorless to light yellow liquid with a density of 0.884 g/cm³. For experimental purposes, it can be dissolved in DMSO and prepared as stock solutions, which should be aliquoted and stored to prevent inactivation from repeated freeze-thaw cycles. Various in vivo dissolution protocols are available, utilizing co-solvents such as PEG300, Tween-80, Saline, SBE-β-CD, or Corn Oil, to achieve clear solutions for different administration routes and experimental animals.
  • Tertiary amine compound.
  • Exhibits anticonvulsant activity.
  • Provided as a liquid for ease of use.
  • Detailed solubility and stock solution preparation guidance is available.
  • Multiple in vivo dissolution protocols are provided to support various experimental designs.
Catalog No. 50-004-51859 Supplier Medchemexpress LLC Supplier No. HYW010383100G
3-Diethylamino-1-propanol is a tertiary amine compound that demonstrates anticonvulsant activity. It is intended for research use only. The product is supplied as a colorless to light yellow liquid with a density of 0.884 g/cm³. For experimental purposes, it can be dissolved in DMSO and prepared as stock solutions, which should be aliquoted and stored to prevent inactivation from repeated freeze-thaw cycles. Various in vivo dissolution protocols are available, utilizing co-solvents such as PEG300, Tween-80, Saline, SBE-β-CD, or Corn Oil, to achieve clear solutions for different administration routes and experimental animals.
  • Tertiary amine compound.
  • Exhibits anticonvulsant activity.
  • Provided as a liquid for ease of use.
  • Detailed solubility and stock solution preparation guidance is available.
  • Multiple in vivo dissolution protocols are provided to support various experimental designs.