Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Diethylamino-1-propanol | 622-93-5 | 98.0% | 131.22 | 25 G
SDP

Catalog No. 5000451496
Encompass_Preferred

3-Diethylamino-1-propanol is a tertiary amine compound that exhibits anticonvulsant activity. It is intended for research use only and is not sold to patients. This liquid appears colorless to light yellow with a density of 0.884 g/cm³.

  • Exhibits anticonvulsant activity
  • Suitable for research applications
  • Liquid form with a colorless to light yellow appearance
  • Molecular formula of C7H17NO
  • Purity of 98.0%
  • Molecular weight of 131.22
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-51496 Supplier Medchemexpress LLC Supplier No. 25GHYW010383
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Diethylamino-1-propanol is a tertiary amine compound that exhibits anticonvulsant activity. It is intended for research use only and is not sold to patients. This liquid appears colorless to light yellow with a density of 0.884 g/cm³.

  • Exhibits anticonvulsant activity
  • Suitable for research applications
  • Liquid form with a colorless to light yellow appearance
  • Molecular formula of C7H17NO
  • Purity of 98.0%
  • Molecular weight of 131.22
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.